Gene Transcripts

Gene Symbol
Txlna
Entrez Gene ID
682457
Gene Name
taxilin alpha
Gene Aliases
Chromosome Location
Chr. 5: 147830514 - 147846196 on Build Rnor_6.0
UniGene ID
Rn.217158
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127633.1 NP_001121105.1 3 363
XM_006238972.2 XP_006239034.1 3 370
GenBank mRNA BC166486.1 - 3 363

View Full Product Details