Gene Transcripts

Gene Symbol
Hist2h3c2
Entrez Gene ID
310678
Gene Name
histone cluster 2, H3c2
Gene Aliases
Chromosome Location
Chr. 2: 198390109 - 198418075 on Build Rnor_6.0
UniGene ID
Rn.218658
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107698.1 NP_001101168.1 1 229
XM_008761309.1 XP_008759531.1 1 286

Gene Symbol
LOC100910366
Entrez Gene ID
100910366
Gene Name
uncharacterized LOC100910366
Gene Aliases
Chromosome Location
Chr. 17: 43791589 - 43802084 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008771686.1 XP_008769908.1 1 297

Gene Symbol
LOC680498
Entrez Gene ID
680498
Gene Name
similar to CG31613-PA
Gene Aliases
Chromosome Location
Chr. 17: 44759999 - 44764062 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001057443.4 XP_001057443.3 2 853

Gene Symbol
LOC690126
Entrez Gene ID
690126
Gene Name
hypothetical protein LOC690126
Gene Aliases
Chromosome Location
Chr. 2: 198379864 - 198380888 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008761346.1 XP_008759568.1 1 281

View Full Product Details