Gene Transcripts

Gene Symbol
Rps28
Entrez Gene ID
691531
Gene Name
ribosomal protein S28
Gene Aliases
Chromosome Location
Chr. 7: 18682071 - 18683440 on Build Rnor_6.0
UniGene ID
Rn.6005
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105730.1 NP_001099200.1 3 130
GenBank mRNA FQ210333.1 - 3 122
FQ211223.1 - 3 122
FQ217403.1 - 3 127
FQ218290.1 - 3 121
FQ221403.1 - 3 122
FQ221642.1 - 3 122
FQ221735.1 - 3 123
FQ222008.1 - 3 122
FQ222172.1 - 3 122
FQ222320.1 - 3 122
FQ222499.1 - 3 122
FQ222594.1 - 3 125
FQ222598.1 - 3 123
FQ222664.1 - 3 122
FQ223680.1 - 3 123
FQ224357.1 - 3 122
FQ228386.1 - 3 123
FQ228826.1 - 3 121
FQ228989.1 - 3 122
FQ229601.1 - 3 122
X59277.1 - 3 111

Gene Symbol
LOC100359503
Entrez Gene ID
100359503
Gene Name
ribosomal protein S28-like
Gene Aliases
Chromosome Location
Chr. 16: 70644447 - 70644750 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002728428.3 XP_002728474.1 1 127

View Full Product Details