Gene Transcripts

Gene Symbol
MORC3
Entrez Gene ID
23515
Gene Name
MORC family CW-type zinc finger 3
Gene Aliases
NXP2, ZCW5, ZCWCC3
Chromosome Location
Chr. 21: 36320189 - 36376646 on Build GRCh38
UniGene ID
Hs.421150
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320445.1 NP_001307374.1 13 1557
NM_001320446.1 NP_001307375.1 15 1716
NM_015358.2 NP_056173.1 14 1630
XM_011529518.2 XP_011527820.1 14 1850
XM_017028311.1 XP_016883800.1 14 1652
XM_017028312.1 XP_016883801.1 13 1475
GenBank mRNA AK292957.1 - 14 1581
AK295133.1 - 13 1549
BC007311.1 - 14 1587
BC094779.1 - 14 1587 30527767
BC132731.1 - 14 1584 40146753
BC144116.1 - 15 1670
D50926.2 - 14 1589

View Full Product Details