Gene Transcripts
- Gene Symbol
- DAPK2
- Entrez Gene ID
- 23604
- Gene Name
- death associated protein kinase 2
- Gene Aliases
- DRP-1, DRP1
- Chromosome Location
- Chr. 15: 63907036 - 64046470 on Build GRCh38
- UniGene ID
- Hs.237886
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014326.3 |
NP_055141.2
|
5, 6 |
611 |
|
XM_011521413.2 |
XP_011519715.1
|
4, 5 |
586 |
|
XM_011521414.1 |
XP_011519716.1
|
4, 5 |
586 |
|
XM_011521415.2 |
XP_011519717.1
|
4, 5 |
586 |
|
XM_011521416.2 |
XP_011519718.1
|
5, 6 |
759 |
|
XM_011521417.1 |
XP_011519719.1
|
4, 5 |
586 |
|
XM_011521421.2 |
XP_011519723.1
|
3, 4 |
396 |
|
XM_017022043.1 |
XP_016877532.1
|
3, 4 |
355 |
|
XM_017022044.1 |
XP_016877533.1
|
4, 5 |
586 |
|
XM_017022045.1 |
XP_016877534.1
|
4, 5 |
586 |
|
XM_017022046.1 |
XP_016877535.1
|
4, 5 |
586 |
|
XM_017022047.1 |
XP_016877536.1
|
3, 4 |
396 |
|
XM_017022048.1 |
XP_016877537.1
|
2, 3 |
345 |
|
XM_017022049.1 |
XP_016877538.1
|
3, 4 |
392 |
|
XM_017022050.1 |
XP_016877539.1
|
2, 3 |
197 |
|
XM_017022051.1 |
XP_016877540.1
|
2, 3 |
343 |
|
GenBank mRNA |
AB018001.1 |
- |
5, 6 |
610 |
|
AF052941.1 |
- |
5, 6 |
611 |
|
AK310967.1 |
- |
6, 7 |
698 |
|
BC114506.1 |
- |
4, 5 |
600 |
40005390 |
BC114951.1 |
- |
4, 5 |
508 |
40005391 |
GU056176.1 |
- |
5, 6 |
611 |
|
View Full Product Details