Gene Transcripts

Gene Symbol
MKRN1
Entrez Gene ID
23608
Gene Name
makorin ring finger protein 1
Gene Aliases
RNF61
Chromosome Location
Chr. 7: 140453040 - 140479569 on Build GRCh38
UniGene ID
Hs.744883
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145125.1 NP_001138597.1 5 1084
NM_001291663.1 NP_001278592.1 7 1246
NM_013446.3 NP_038474.2 5 1084
NR_117084.1 3 627
XM_011515996.1 XP_011514298.1 4 873
XM_011515997.2 XP_011514299.1 5 1141
XM_011515998.1 XP_011514300.1 5 826
GenBank mRNA AB209298.1 - 5 851
AF117233.1 - 5 1011
AF192784.1 - 5 979
AK027137.1 - 4 596
AK127030.1 - 6 1701
AK297361.1 - 3 426
AK315552.1 - 5 883
AL136812.1 - 5 982
AM236048.1 - 5 865
BC025955.1 - 5 1017 4869484
BC037400.1 - 5 869 5176176
BC064838.1 - 5 862 6173715

Gene Symbol
LOC441455
Entrez Gene ID
441455
Gene Name
makorin ring finger protein 1 pseudogene
Gene Aliases
Chromosome Location
Chr. 9: 96725821 - 96727467 on Build GRCh38
UniGene ID
Hs.229613
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026792.1 1 838

View Full Product Details