Gene Transcripts

Gene Symbol
RASGRP3
Entrez Gene ID
25780
Gene Name
RAS guanyl releasing protein 3
Gene Aliases
GRP3
Chromosome Location
Chr. 2: 33436348 - 33564731 on Build GRCh38
UniGene ID
Hs.143674
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001139488.1 NP_001132960.1 10 1629
NM_015376.2 NP_056191.1 9 1187
NM_170672.2 NP_733772.1 11 1542
XM_005264246.3 XP_005264303.1 10 1969
XM_011532746.2 XP_011531048.1 11 1416
XM_011532747.2 XP_011531049.1 10 1431
XM_011532748.2 XP_011531050.2 10 1277
XM_017003759.1 XP_016859248.1 12 2892
XM_017003760.1 XP_016859249.1 11 3364
XM_017003761.1 XP_016859250.1 10 1294
XM_017003762.1 XP_016859251.1 11 1543
XM_017003763.1 XP_016859252.1 10 1273
GenBank mRNA AB020653.1 - 9 1187
AK292983.1 - 11 1453
BC027849.1 - 11 1542 5203041
BX647990.1 - 10 1237

View Full Product Details