Gene Transcripts

Gene Symbol
METTL7A
Entrez Gene ID
25840
Gene Name
methyltransferase like 7A
Gene Aliases
AAM-B
Chromosome Location
Chr. 12: 50923811 - 50932517 on Build GRCh38
UniGene ID
Hs.711113
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014033.3 NP_054752.3 1 421
XM_006719332.1 XP_006719395.1 2 775
GenBank mRNA AF113007.1 - 1 95
AJ536233.1 - 1 143
AK023693.1 - 1 153
AK024409.1 - 1 186
AK225595.1 - 1 158
AK225689.1 - 1 157
AK293356.1 - 1 154
AK307071.1 - 1 161
AL050159.1 - 1 131
AY358797.1 - 1 189
BC004492.2 - 1 138 3689692
BC008180.2 - 1 139 2900478
BM805242.1 - 1 443

View Full Product Details