Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015629.3 | NP_056444.3 | 5 | 739 | |
XM_006723137.3 | XP_006723200.1 | 5 | 649 | ||
GenBank mRNA | AB593024.1 | - | 6 | 1658 | |
AB593025.1 | - | 5 | 395 | ||
AF308303.1 | - | 4 | 214 | ||
AK098547.1 | - | 5 | 378 | ||
AK301709.1 | - | 5 | 404 | ||
AL050369.1 | - | 5 | 378 | ||
AY040822.1 | - | 5 | 700 | ||
BC117389.1 | - | 5 | 663 | 40125940 |