Gene Transcripts
- Gene Symbol
- FBXO4
- Entrez Gene ID
- 26272
- Gene Name
- F-box protein 4
- Gene Aliases
- FBX4
- Chromosome Location
- Chr. 5: 41925249 - 42040230 on Build GRCh38
- UniGene ID
- Hs.165575
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001297437.1 |
NP_001284366.1
|
3, 4 |
702 |
|
NM_012176.2 |
NP_036308.1
|
3, 4 |
700 |
|
NM_033484.2 |
NP_277019.1
|
3, 4 |
700 |
|
XM_006714470.3 |
XP_006714533.1
|
2, 3 |
398 |
|
XM_011514026.2 |
XP_011512328.1
|
3, 4 |
705 |
|
XM_011514027.2 |
XP_011512329.1
|
3, 4 |
2075 |
|
GenBank mRNA |
AF129534.1 |
- |
3, 4 |
644 |
|
AF176703.1 |
- |
3, 4 |
638 |
|
AK024075.1 |
- |
3, 4 |
700 |
|
AU143782.1 |
- |
3, 4 |
700 |
|
BC040011.1 |
- |
3, 4 |
661 |
|
BC048098.1 |
- |
3, 4 |
668 |
4830192 |
CR749719.1 |
- |
1, 2 |
217 |
|
View Full Product Details