Gene Transcripts

Gene Symbol
OR4E2
Entrez Gene ID
26686
Gene Name
olfactory receptor family 4 subfamily E member 2
Gene Aliases
OR14-42
Chromosome Location
Chr. 14: 21665083 - 21666024 on Build GRCh38
UniGene ID
Hs.547982
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001912.2 NP_001001912.2 1 243
GenBank mRNA AY726598.1 - 1 191

View Full Product Details