Gene Transcripts
- Gene Symbol
- DKKL1
- Entrez Gene ID
- 27120
- Gene Name
- dickkopf like acrosomal protein 1
- Gene Aliases
- CT34, SGY, SGY-1, SGY1
- Chromosome Location
- Chr. 19: 49360520 - 49375132 on Build GRCh38
- UniGene ID
- Hs.515855
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001197302.1 |
NP_001184231.1
|
4, 5 |
585 |
|
NM_014419.3 |
NP_055234.1
|
4, 5 |
624 |
|
XM_006723142.2 |
XP_006723205.1
|
4, 5 |
470 |
|
XM_011526724.1 |
XP_011525026.1
|
4, 5 |
494 |
|
XM_011526725.1 |
XP_011525027.1
|
4, 5 |
444 |
|
XM_011526726.2 |
XP_011525028.1
|
4, 5 |
704 |
|
XM_011526729.1 |
XP_011525031.1
|
4, 5 |
465 |
|
XM_017026593.1 |
XP_016882082.1
|
4, 5 |
477 |
|
GenBank mRNA |
AF177398.1 |
- |
4, 5 |
475 |
|
AY358922.1 |
- |
4, 5 |
624 |
|
BC030581.2 |
- |
4, 5 |
504 |
4836991 |
BI826695.1 |
- |
4, 5 |
585 |
|
View Full Product Details