Gene Transcripts

Gene Symbol
SULT1C4
Entrez Gene ID
27233
Gene Name
sulfotransferase family 1C member 4
Gene Aliases
SULT1C, SULT1C2
Chromosome Location
Chr. 2: 108377911 - 108388989 on Build GRCh38
UniGene ID
Hs.312644
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321770.1 NP_001308699.1 2 657
NM_006588.3 NP_006579.2 2 657
NR_135776.1 2 707
XM_017003807.1 XP_016859296.1 2 199
GenBank mRNA AK297851.1 - 2 435
AK298755.1 - 2 598
BC058861.1 - 2 657
BC125043.1 - 2 383 40115974
HY118496.1 - 1 74

View Full Product Details