Gene Transcripts

Gene Symbol
UQCR10
Entrez Gene ID
29796
Gene Name
ubiquinol-cytochrome c reductase, complex III subunit X
Gene Aliases
HSPC051, HSPC119, HSPC151, QCR9, UCCR7.2, UCRC
Chromosome Location
Chr. 22: 29767369 - 29770413 on Build GRCh38
UniGene ID
Hs.284292
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003684.1 NP_001003684.1 2 684
NM_013387.3 NP_037519.2 2 625
GenBank mRNA BI754905.1 - 2 587
CD674029.1 - 2 492

View Full Product Details