Gene Transcripts

Gene Symbol
OSGIN1
Entrez Gene ID
29948
Gene Name
oxidative stress induced growth inhibitor 1
Gene Aliases
BDGI, OKL38
Chromosome Location
Chr. 16: 83953222 - 83966332 on Build GRCh38
UniGene ID
Hs.128055
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182981.2 NP_892026.1 2, 3 242
GenBank mRNA AK290383.1 - 2, 3 224
AY037158.1 - 2, 3 207
AY258066.1 - 3, 4 693
AY258067.1 - 2, 3 533
AY258068.1 - 2, 3 223
BC020434.1 - 2, 3 208
BC032476.1 - 2, 3 207 5215262
BC093687.1 - 2, 3 125 7939532
BC113417.1 - 2, 3 125 8322469

View Full Product Details