Gene Transcripts
- Gene Symbol
- CD48
- Entrez Gene ID
- 962
- Gene Name
- CD48 molecule
- Gene Aliases
- BCM1, BLAST, BLAST1, MEM-102, SLAMF2, hCD48, mCD48
- Chromosome Location
- Chr. 1: 160678746 - 160711851 on Build GRCh38
- UniGene ID
- Hs.243564
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001256030.1 |
NP_001242959.1
|
1, 2 |
175 |
|
NM_001778.3 |
NP_001769.2
|
1, 2 |
175 |
|
XM_005245625.1 |
XP_005245682.1
|
1, 2 |
146 |
|
GenBank mRNA |
AK225636.1 |
- |
1, 2 |
148 |
|
AK310624.1 |
- |
1, 2 |
149 |
|
AK310744.1 |
- |
1, 2 |
155 |
|
BC016182.1 |
- |
1, 2 |
119 |
4850933 |
BC030224.1 |
- |
1, 2 |
127 |
5223308 |
BG505217.1 |
- |
1, 2 |
175 |
|
BT019813.1 |
- |
1, 2 |
87 |
|
CR457012.1 |
- |
1, 2 |
87 |
|
M37766.1 |
- |
1, 2 |
107 |
|
M59904.1 |
- |
1, 2 |
104 |
|
X06341.1 |
- |
1, 2 |
123 |
|
View Full Product Details