Gene Transcripts

Gene Symbol
LRBA
Entrez Gene ID
987
Gene Name
LPS responsive beige-like anchor protein
Gene Aliases
BGL, CDC4L, CVID8, LAB300, LBA
Chromosome Location
Chr. 4: 150264515 - 151015725 on Build GRCh38
UniGene ID
Hs.480938
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199282.2 NP_001186211.2 28 4776
NM_006726.4 NP_006717.2 28 4735
XM_005263372.3 XP_005263429.1 28 4921
XM_005263373.2 XP_005263430.1 28 4963
XM_005263374.3 XP_005263431.1 28 4920
XM_005263375.3 XP_005263432.1 28 4920
XM_011532434.2 XP_011530736.1 28 4920
XM_017008872.1 XP_016864361.1 28 4963
GenBank mRNA AB208838.1 - 4 449
AF216648.2 - 28 4735
AF467287.1 - 28 4776
M83822.1 - 10 2171

View Full Product Details