Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016120.3 | NP_057204.2 | 4 | 770 | |
NM_183353.2 | NP_899196.1 | 5 | 832 | ||
GenBank mRNA | AF155109.1 | - | 4 | 722 | |
AK001334.1 | - | 4 | 752 | ||
AK314760.1 | - | 1 | 326 | ||
BC013357.2 | - | 4 | 734 | 3834870 |