Gene Transcripts
- Gene Symbol
- WAC
- Entrez Gene ID
- 51322
- Gene Name
- WW domain containing adaptor with coiled-coil
- Gene Aliases
- BM-016, DESSH, PRO1741, Wwp4
- Chromosome Location
- Chr. 10: 28532426 - 28623112 on Build GRCh38
- UniGene ID
- Hs.743224
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016628.4 |
NP_057712.2
|
2, 3 |
175 |
|
NM_100264.2 |
NP_567822.1
|
2, 3 |
466 |
|
NM_100486.3 |
NP_567823.1
|
2, 3 |
175 |
|
XM_011519491.2 |
XP_011517793.1
|
2, 3 |
776 |
|
XM_017016315.1 |
XP_016871804.1
|
2, 3 |
286 |
|
XM_017016316.1 |
XP_016871805.1
|
2, 3 |
776 |
|
XM_017016317.1 |
XP_016871806.1
|
2, 3 |
286 |
|
XM_017016318.1 |
XP_016871807.1
|
2, 3 |
286 |
|
GenBank mRNA |
AF116666.2 |
- |
3, 4 |
240 |
|
AK055852.1 |
- |
2, 3 |
283 |
|
AK223600.1 |
- |
2, 3 |
173 |
|
AK225846.1 |
- |
2, 3 |
279 |
|
AK290174.1 |
- |
2, 3 |
180 |
|
AK297460.1 |
- |
3 |
88 |
|
AL713727.1 |
- |
1 |
305 |
|
BC004258.1 |
- |
2, 3 |
166 |
3347345 |
BI226998.1 |
- |
2, 3 |
63 |
|
BI463901.1 |
- |
2, 3 |
566 |
|
CB143386.1 |
- |
2, 3 |
354 |
|
View Full Product Details