Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001111319.1 | NP_001104789.1 | 1 | 755 | |
GenBank mRNA | AK098064.1 | - | 1 | 149 | |
AK303864.1 | - | 249 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001185149.1 | NP_001172078.1 | 1 | 199 |