Gene Transcripts
- Gene Symbol
- TEX10
- Entrez Gene ID
- 54881
- Gene Name
- testis expressed 10
- Gene Aliases
- Ipi1, bA208F1.2
- Chromosome Location
- Chr. 9: 100302075 - 100352977 on Build GRCh38
- UniGene ID
- Hs.494648
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001161584.1 |
NP_001155056.1
|
8, 9 |
1958 |
|
NM_017746.3 |
NP_060216.2
|
8, 9 |
2014 |
|
XM_011518798.1 |
XP_011517100.1
|
6, 7 |
1601 |
|
XM_017014848.1 |
XP_016870337.1
|
8, 9 |
1503 |
|
GenBank mRNA |
AB060968.1 |
- |
8, 9 |
1930 |
|
AJ420436.1 |
- |
3, 4 |
439 |
|
AK000294.1 |
- |
8, 9 |
1930 |
|
AK298916.1 |
- |
10, 11 |
1537 |
|
AK302619.1 |
- |
8, 9 |
1961 |
|
AY598337.1 |
- |
7, 8 |
1799 |
|
BC030652.2 |
- |
8, 9 |
1850 |
5268105 |
View Full Product Details