Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317177.1 | NP_001304106.1 | 2 | 370 | |
NM_001317178.1 | NP_001304107.1 | 2 | 370 | ||
NM_017812.3 | NP_060282.1 | 2 | 370 | ||
NR_133671.1 | 2 | 370 | |||
GenBank mRNA | AK000427.1 | - | 2 | 286 | |
AK129533.1 | - | 2 | 302 | ||
AK294071.1 | - | 2 | 252 | ||
AK310236.1 | - | 2 | 282 | ||
AL516679.3 | - | 2 | 267 | ||
BC011596.2 | - | 2 | 292 | 3346346 | |
BC014839.1 | - | 2 | 294 | 2901125 | |
DC385940.1 | - | 2 | 371 |