Gene Transcripts

Gene Symbol
ANKZF1
Entrez Gene ID
55139
Gene Name
ankyrin repeat and zinc finger domain containing 1
Gene Aliases
ZNF744
Chromosome Location
Chr. 2: 219229757 - 219236669 on Build GRCh38
UniGene ID
Hs.437647
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042410.1 NP_001035869.1 11 1835
NM_001282792.1 NP_001269721.1 8 1514
NM_018089.2 NP_060559.2 11 1908
XM_005246663.2 XP_005246720.1 11 1827
XM_011511392.2 XP_011509694.1 11 1905
XM_017004420.1 XP_016859909.1 11 1824
GenBank mRNA AF364318.1 - 10 2298
AK001277.1 - 11 1844
AK023206.1 - 11 1835
AK056060.1 - 1 1220
AK095304.1 - 9 2470
AK225275.1 - 11 1833
AK225309.1 - 11 1809
BC000238.1 - 11 1790 3351742
BC008948.2 - 11 1819 2959748
BC028334.1 - 8 1436
DQ412564.1 - 11 1844

View Full Product Details