Gene Transcripts

Gene Symbol
SLC22A11
Entrez Gene ID
55867
Gene Name
solute carrier family 22 member 11
Gene Aliases
OAT4, hOAT4
Chromosome Location
Chr. 11: 64555601 - 64572875 on Build GRCh38
UniGene ID
Hs.220844
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018484.3 NP_060954.1 6, 7 1468
XM_011545167.1 XP_011543469.1 5, 6 1118
GenBank mRNA AB026116.1 - 6, 7 1121
AK075224.1 - 6, 7 814
AK222869.1 - 6, 7 1126
AK290791.1 - 6, 7 1451
AK309931.1 - 6, 7 1128
BC034384.1 - 6, 7 1443 5190509

View Full Product Details