Gene Transcripts
- Gene Symbol
- CNN1
- Entrez Gene ID
- 1264
- Gene Name
- calponin 1
- Gene Aliases
- HEL-S-14, SMCC, Sm-Calp
- Chromosome Location
- Chr. 19: 11538717 - 11550323 on Build GRCh38
- UniGene ID
- Hs.465929
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001299.5 |
NP_001290.2
|
2, 3 |
389 |
|
NM_001308341.1 |
NP_001295270.1
|
3, 4 |
664 |
|
NM_001308342.1 |
NP_001295271.1
|
3, 4 |
655 |
|
XM_005259741.2 |
XP_005259798.1
|
2, 3 |
238 |
|
XM_017026289.1 |
XP_016881778.1
|
2, 3 |
561 |
|
GenBank mRNA |
AK223234.1 |
- |
2, 3 |
255 |
|
AK300837.1 |
- |
2, 3 |
169 |
|
AK301888.1 |
- |
3, 4 |
528 |
|
AK313255.1 |
- |
2, 3 |
258 |
|
AK316406.1 |
- |
3, 4 |
605 |
|
BC022015.2 |
- |
2, 3 |
342 |
4824832 |
BC036307.1 |
- |
2, 3 |
328 |
5165618 |
BG718473.1 |
- |
2, 3 |
392 |
|
D17408.1 |
- |
2, 3 |
270 |
|
EU794601.1 |
- |
2, 3 |
328 |
|
S80560.1 |
- |
2, 3 |
252 |
|
U37019.1 |
- |
2, 3 |
256 |
|
View Full Product Details