Gene Transcripts

Gene Symbol
PLEKHH1
Entrez Gene ID
57475
Gene Name
pleckstrin homology, MyTH4 and FERM domain containing H1
Gene Aliases
Chromosome Location
Chr. 14: 67533155 - 67589766 on Build GRCh38
UniGene ID
Hs.594236
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020715.2 NP_065766.1 7 1178
XM_011537009.1 XP_011535311.1 7 1270
XM_017021499.1 XP_016876988.1 6 1251
XM_017021500.1 XP_016876989.1 7 1270
XM_017021501.1 XP_016876990.1 6 1251
XM_017021503.1 XP_016876992.1 1 486
XM_017021504.1 XP_016876993.1 3 712
GenBank mRNA AI792090.1 - 1 208
AK307428.1 - 7 1270

View Full Product Details