Gene Transcripts

Gene Symbol
SHROOM4
Entrez Gene ID
57477
Gene Name
shroom family member 4
Gene Aliases
SHAP
Chromosome Location
Chr. X: 50575534 - 50814167 on Build GRCh38
UniGene ID
Hs.420541
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020717.3 NP_065768.2 4 2072
NR_027121.1 4 2072
XM_017029682.1 XP_016885171.1 4 2178
XM_017029683.1 XP_016885172.1 3 2043
XM_017029684.1 XP_016885173.1 3 2195
XM_017029685.1 XP_016885174.1 4 2195
XM_017029686.1 XP_016885175.1 2 2060
XM_017029687.1 XP_016885176.1 4 2182
GenBank mRNA AB033028.1 - 4 2072
AY044234.1 - 3 2153
BC151240.1 - 4 2083 8860513

View Full Product Details