Gene Transcripts

Gene Symbol
MTA3
Entrez Gene ID
57504
Gene Name
metastasis associated 1 family member 3
Gene Aliases
Chromosome Location
Chr. 2: 42494569 - 42795032 on Build GRCh38
UniGene ID
Hs.435413
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282755.1 NP_001269684.1 3 584
NM_001282756.1 NP_001269685.1 2 236
NM_020744.3 NP_065795.1 2 311
XM_005264456.2 XP_005264513.1 2 113
XM_005264458.2 XP_005264515.1 2 113
XM_005264459.2 XP_005264516.1 3 584
XM_017004561.1 XP_016860050.1 2 113
XM_017004562.1 XP_016860051.1 2 113
GenBank mRNA AB033092.1 - 2 236
AK027304.1 - 2 109
AK127245.1 - 3 584
BC004227.2 - 2 139 3542706
BC053631.1 - 2 297 6472900
BU859486.1 - 2 311

View Full Product Details