Gene Transcripts

Gene Symbol
ZBTB2
Entrez Gene ID
57621
Gene Name
zinc finger and BTB domain containing 2
Gene Aliases
ZNF437
Chromosome Location
Chr. 6: 151364115 - 151391705 on Build GRCh38
UniGene ID
Hs.520073
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020861.2 NP_065912.1 3 1677
XM_005267076.2 XP_005267133.1 3 1538
XM_011536004.2 XP_011534306.1 3 1759
XM_017011137.1 XP_016866626.1 4 1937
GenBank mRNA AB040916.1 - 1 1128
AJ420526.1 - 1 913
AK092886.1 - 4 1443
AK291942.1 - 3 1620
AL832957.1 - 1 1449
BC020172.1 - 3 1519 4577244

View Full Product Details