Gene Transcripts

Gene Symbol
GRHL3
Entrez Gene ID
57822
Gene Name
grainyhead like transcription factor 3
Gene Aliases
SOM, TFCP2L4, VWS2
Chromosome Location
Chr. 1: 24319322 - 24364482 on Build GRCh38
UniGene ID
Hs.657920
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195010.1 NP_001181939.1 15, 16 1867
NM_021180.3 NP_067003.2 15, 16 1763
NM_198173.2 NP_937816.1 15, 16 1932
XM_011541869.1 XP_011540171.1 15, 16 1813
XM_011541870.2 XP_011540172.1 14, 15 1734
GenBank mRNA AK315164.1 - 15, 16 1734
AL137763.1 - 3, 4 248
AY231160.1 - 15, 16 1763
AY231161.1 - 14, 15 1576
BC036890.2 - 15, 16 1878 5502632

View Full Product Details