Gene Transcripts

Gene Symbol
GAL3ST2
Entrez Gene ID
64090
Gene Name
galactose-3-O-sulfotransferase 2
Gene Aliases
GAL3ST-2, GP3ST
Chromosome Location
Chr. 2: 241776825 - 241804287 on Build GRCh38
UniGene ID
Hs.632556
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022134.2 NP_071417.2 4 1126
GenBank mRNA AB040610.1 - 4 1127
BC117293.1 - 4 1044 40125844
BC117295.1 - 4 1044 40125846

View Full Product Details