Gene Transcripts

Gene Symbol
TNMD
Entrez Gene ID
64102
Gene Name
tenomodulin
Gene Aliases
BRICD4, CHM1L, TEM
Chromosome Location
Chr. X: 100584793 - 100599885 on Build GRCh38
UniGene ID
Hs.132957
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022144.2 NP_071427.2 5 792
GenBank mRNA AB055421.1 - 5 572
AF191770.1 - 5 752
AF234259.1 - 5 633
AF291656.1 - 5 638
AY358706.1 - 5 624

View Full Product Details