Gene Transcripts
- Gene Symbol
- SIL1
- Entrez Gene ID
- 64374
- Gene Name
- SIL1 nucleotide exchange factor
- Gene Aliases
- BAP, MSS, ULG5
- Chromosome Location
- Chr. 5: 138946720 - 139198376 on Build GRCh38
- UniGene ID
- Hs.483521
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001037633.1 |
NP_001032722.1
|
5, 6 |
561 |
|
NM_022464.4 |
NP_071909.1
|
4, 5 |
473 |
|
XM_011543570.2 |
XP_011541872.1
|
6, 7 |
598 |
|
GenBank mRNA |
AF547994.1 |
- |
4, 5 |
392 |
|
AJ299442.1 |
- |
4, 5 |
451 |
|
AK074624.1 |
- |
4, 5 |
465 |
|
AK075177.1 |
- |
4, 5 |
402 |
|
AY358950.1 |
- |
4, 5 |
419 |
|
BC011568.1 |
- |
4, 5 |
441 |
4640182 |
BI561751.1 |
- |
4, 5 |
504 |
|
CB125000.1 |
- |
4, 5 |
473 |
|
KU178743.1 |
- |
3, 4 |
355 |
|
KU178744.1 |
- |
3, 4 |
355 |
|
View Full Product Details