Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032476.3 | NP_115865.1 | 3 | 426 | |
GenBank mRNA | AB049942.1 | - | 3 | 372 | |
AK096374.1 | - | 3 | 1393 | ||
AK312084.1 | - | 3 | 371 | ||
AY061855.1 | - | 3 | 426 | ||
BC000547.2 | - | 3 | 358 | 3162515 | |
BC004976.1 | - | 3 | 360 | 2958115 | |
BC010076.2 | - | 3 | 360 | 4304420 | |
BC051302.1 | - | 3 | 267 | 4855355 | |
BG720377.1 | - | 3 | 376 |