Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320464.1 | NP_001307393.1 | 11 | 1606 | |
NM_024031.3 | NP_076936.1 | 11 | 1762 | ||
GenBank mRNA | AK074783.1 | - | 11 | 1617 | |
AK125716.1 | - | 9 | 1709 | ||
BC000119.1 | - | 11 | 1629 | 3351963 | |
BC010232.1 | - | 9 | 1371 | ||
BC021934.1 | - | 10 | 1714 | 4876482 | |
BC050677.1 | - | 11 | 1709 | 6053861 | |
CR936781.1 | - | 9 | 1220 |