Gene Transcripts
- Gene Symbol
- CHCHD7
- Entrez Gene ID
- 79145
- Gene Name
- coiled-coil-helix-coiled-coil-helix domain containing 7
- Gene Aliases
- COX23
- Chromosome Location
- Chr. 8: 56211638 - 56218798 on Build GRCh38
- UniGene ID
- Hs.436913
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001011667.2 |
NP_001011667.1
|
4, 5 |
568 |
|
NM_001011668.2 |
NP_001011668.1
|
4, 5 |
524 |
|
NM_001011669.2 |
NP_001011669.1
|
4, 5 |
548 |
|
NM_001011670.2 |
NP_001011670.1
|
3, 4 |
420 |
|
NM_001011671.2 |
NP_001011671.1
|
3, 4 |
376 |
|
NM_024300.4 |
NP_077276.2
|
4, 5 |
504 |
|
NR_133937.1 |
|
3, 4 |
416 |
|
GenBank mRNA |
AK095922.1 |
- |
3, 4 |
225 |
|
AK098285.1 |
- |
2, 3 |
3027 |
|
AK225982.1 |
- |
4, 5 |
363 |
|
AK290088.1 |
- |
4, 5 |
353 |
|
AY070434.1 |
- |
4, 5 |
396 |
|
BC002546.2 |
- |
4, 5 |
343 |
3139026 |
BM695222.1 |
- |
3, 4 |
260 |
|
BX473986.1 |
- |
4, 5 |
406 |
|
View Full Product Details