Gene Transcripts

Gene Symbol
GCC1
Entrez Gene ID
79571
Gene Name
GRIP and coiled-coil domain containing 1
Gene Aliases
GCC1P, GCC88
Chromosome Location
Chr. 7: 127580628 - 127585600 on Build GRCh38
UniGene ID
Hs.521168
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024523.5 NP_078799.2 2 2701
GenBank mRNA AF525417.1 - 2 2476
AK025688.1 - 2 1360
AK092878.1 - 2 2157
AK315472.1 - 2 2552
AL832833.1 - 2 1363
BC008902.2 - 2 1483
BC014100.2 - 2 2701 2988591
BC078665.1 - 2 2653 5923117

View Full Product Details