Gene Transcripts

Gene Symbol
C1orf54
Entrez Gene ID
79630
Gene Name
chromosome 1 open reading frame 54
Gene Aliases
Chromosome Location
Chr. 1: 150272284 - 150280927 on Build GRCh38
UniGene ID
Hs.91283
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301039.1 NP_001287968.1 4 307
NM_001301040.1 NP_001287969.1 4 307
NM_001301041.1 NP_001287970.1 2 93
NM_001301042.1 NP_001287971.1 3 177
NM_024579.3 NP_078855.2 3 196
XM_011509984.2 XP_011508286.1 3 223
GenBank mRNA AA095731.1 - 3 164
AK026874.1 - 3 181
AX770997.1 - 3 194
AX770998.1 - 4 221
AX770999.1 - 3 196
BC017761.1 - 3 181 4617936
DA358536.1 - 2 104

View Full Product Details