Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001018089.2 | NP_001018099.1 | 10 | 1941 | |
NM_024611.5 | NP_078887.2 | 10 | 2101 | ||
XM_005254661.1 | XP_005254718.1 | 11 | 2100 | ||
XM_005254662.1 | XP_005254719.1 | 11 | 1963 | ||
XM_011522009.1 | XP_011520311.1 | 10 | 1975 | ||
XM_011522010.1 | XP_011520312.1 | 10 | 1844 | ||
XM_011522011.2 | XP_011520313.1 | 9 | 1758 | ||
XM_017022569.1 | XP_016878058.1 | 11 | 2137 | ||
GenBank mRNA | AF502591.1 | - | 10 | 2044 | |
AK021958.1 | - | 1 | 95 | ||
AK055752.1 | - | 10 | 2044 | ||
AK095389.1 | - | 10 | 1925 | ||
AK127892.1 | - | 3 | 926 | ||
AL832046.1 | - | 10 | 2068 | ||
AL833318.1 | - | 2 | 905 | ||
BC013684.1 | - | 1 | 96 | ||
BC107427.1 | - | 1 | 74 | ||
BC140894.1 | - | 10 | 2020 | 9021762 | |
BX647873.1 | - | 10 | 1927 | ||
CR749852.1 | - | 10 | 1894 |