Gene Transcripts

Gene Symbol
LRRK1
Entrez Gene ID
79705
Gene Name
leucine rich repeat kinase 1
Gene Aliases
RIPK6, Roco1
Chromosome Location
Chr. 15: 100919017 - 101070837 on Build GRCh38
UniGene ID
Hs.407918
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024652.4 NP_078928.3 32 5996
XM_005254979.3 XP_005255036.1 18 3654
XM_011522012.2 XP_011520314.1 34 6270
XM_011522013.2 XP_011520315.1 32 5979
XM_011522014.2 XP_011520316.1 32 6194
XM_011522015.2 XP_011520317.1 27 6789
XM_011522016.2 XP_011520318.1 30 5471
XM_011522017.2 XP_011520319.1 18 3661
XM_011522018.2 XP_011520320.1 18 3656
XM_017022570.1 XP_016878059.1 22 4243
GenBank mRNA AB058693.1 - 18 3701
AK026772.1 - 1 371
AK054662.1 - 6 1444
AK058028.1 - 12 2544
AK094556.1 - 2 1124
AK131075.1 - 13 2929
AK225575.1 - 1 371
BC005408.1 - 7 1667
BC020379.1 - 7 1667
BC066655.1 - 3 909
BC110293.1 - 1 555
BC144705.1 - 30 5384
BC146769.1 - 18 3701
BC171802.1 - 31 5863
DQ013130.1 - 31 5634

View Full Product Details