Gene Transcripts

Gene Symbol
MORC4
Entrez Gene ID
79710
Gene Name
MORC family CW-type zinc finger 4
Gene Aliases
ZCW4, ZCWCC2, dJ75H8.2
Chromosome Location
Chr. X: 106940300 - 107000244 on Build GRCh38
UniGene ID
Hs.496544
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085354.2 NP_001078823.1 15 2545
NM_024657.4 NP_078933.3 15 2545
XM_005262190.3 XP_005262247.1 14 2435
XM_006724691.2 XP_006724754.1 13 2179
XM_011531027.2 XP_011529329.1 12 1833
XM_017029844.1 XP_016885333.1 14 2435
GenBank mRNA AK021627.1 - 4 859
AK056235.1 - 11 1757
AK300305.1 - 14 2120
EF125209.1 - 15 2270
EF125210.1 - 14 2329

View Full Product Details