Gene Transcripts
- Gene Symbol
- NAA40
- Entrez Gene ID
- 79829
- Gene Name
- N(alpha)-acetyltransferase 40, NatD catalytic subunit
- Gene Aliases
- NAT11, PATT1
- Chromosome Location
- Chr. 11: 63938970 - 63957322 on Build GRCh38
- UniGene ID
- Hs.523753
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001300800.1 |
NP_001287729.1
|
4, 5 |
442 |
|
NM_024771.3 |
NP_079047.2
|
4, 5 |
371 |
|
XM_006718689.3 |
XP_006718752.1
|
4, 5 |
358 |
|
XM_011545254.2 |
XP_011543556.1
|
5, 6 |
445 |
|
XM_011545255.2 |
XP_011543557.1
|
5, 6 |
445 |
|
GenBank mRNA |
AK023910.1 |
- |
4, 5 |
340 |
|
AK299041.1 |
- |
4, 5 |
436 |
|
AK300451.1 |
- |
3, 4 |
292 |
|
AK303557.1 |
- |
3 |
292 |
|
BC041617.1 |
- |
4, 5 |
300 |
5921480 |
BC052298.1 |
- |
4, 5 |
300 |
6292760 |
BX647309.1 |
- |
3, 4 |
1397 |
|
DA649381.1 |
- |
4, 5 |
376 |
|
DC307984.1 |
- |
4, 5 |
525 |
|
HY009487.1 |
- |
4, 5 |
442 |
|
JF263566.1 |
- |
4, 5 |
340 |
|
View Full Product Details