Gene Transcripts

Gene Symbol
PABPC1L
Entrez Gene ID
80336
Gene Name
poly(A) binding protein cytoplasmic 1 like
Gene Aliases
C20orf119, EPAB, PABPC1L1, dJ1069P2.3
Chromosome Location
Chr. 20: 44910060 - 44959160 on Build GRCh38
UniGene ID
Hs.641481
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001124756.2 NP_001118228.1 7 1001
NR_134983.1 7 1001
GenBank mRNA BX451955.2 - 2 162
EU190483.1 - 7 937

View Full Product Details