Gene Transcripts

Gene Symbol
REEP4
Entrez Gene ID
80346
Gene Name
receptor accessory protein 4
Gene Aliases
C8orf20, PP432, Yip2c
Chromosome Location
Chr. 8: 22138014 - 22141951 on Build GRCh38
UniGene ID
Hs.289063
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316964.1 NP_001303893.1 3 602
NM_001316965.1 NP_001303894.1 3 602
NM_025232.3 NP_079508.2 3 602
GenBank mRNA AF218014.1 - 3 380
AK025899.1 - 3 532
AK025930.1 - 3 235
AK172846.1 - 3 537
AK302352.1 - 3 419
AY562242.1 - 3 133
BC013048.1 - 3 586 3863838
BC050622.1 - 3 504 6504298
BG695963.1 - 3 191
CR457353.1 - 3 133

View Full Product Details