Gene Transcripts
- Gene Symbol
- LY6G6C
- Entrez Gene ID
- 80740
- Gene Name
- lymphocyte antigen 6 complex, locus G6C
- Gene Aliases
- C6orf24, G6c, NG24
- Chromosome Location
- Chr. 6: 31718648 - 31721734 on Build GRCh38
- UniGene ID
- Hs.732272
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC036302.1 |
- |
3 |
283 |
|
View Full Product Details