Gene Transcripts
- Gene Symbol
- VOPP1
- Entrez Gene ID
- 81552
- Gene Name
- vesicular, overexpressed in cancer, prosurvival protein 1
- Gene Aliases
- ECOP, GASP
- Chromosome Location
- Chr. 7: 55434406 - 55572520 on Build GRCh38
- UniGene ID
- Hs.488307
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001321243.1 |
NP_001308172.1
|
1, 2 |
249 |
|
NM_001321246.1 |
NP_001308175.1
|
1, 2 |
249 |
|
NM_001321247.1 |
NP_001308176.1
|
1, 2 |
249 |
|
NM_030796.4 |
NP_110423.3
|
1, 2 |
249 |
|
XM_011515539.1 |
XP_011513841.1
|
1, 2 |
191 |
|
GenBank mRNA |
AB097004.1 |
- |
1, 2 |
108 |
|
AF395824.1 |
- |
|
14 |
|
AK075391.1 |
- |
1, 2 |
183 |
|
AK225755.1 |
- |
1, 2 |
194 |
|
AK314338.1 |
- |
1, 2 |
142 |
|
AL136610.1 |
- |
1 |
62 |
|
BC016650.1 |
- |
1 |
141 |
4342536 |
BI597655.1 |
- |
|
175 |
|
CN299440.1 |
- |
1, 2 |
255 |
|
CR533483.1 |
- |
1, 2 |
53 |
|
DA257111.1 |
- |
1, 2 |
142 |
|
DB475432.1 |
- |
1, 2 |
254 |
|
DB496308.1 |
- |
1, 2 |
254 |
|
HY057980.1 |
- |
1, 2 |
170 |
|
View Full Product Details