Gene Transcripts

Gene Symbol
CALN1
Entrez Gene ID
83698
Gene Name
calneuron 1
Gene Aliases
CABP8
Chromosome Location
Chr. 7: 71779491 - 72504271 on Build GRCh38
UniGene ID
Hs.592750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017440.2 NP_001017440.1 5 934
NM_031468.3 NP_113656.2 6 924
XM_011516594.2 XP_011514896.1 5 776
XM_011516595.1 XP_011514897.1 5 524
XM_011516596.2 XP_011514898.1 6 1419
XM_011516597.2 XP_011514899.1 5 1352
XM_011516599.1 XP_011514901.1 5 723
XM_017012675.1 XP_016868164.1 7 1279
XM_017012676.1 XP_016868165.1 6 1061
XM_017012677.1 XP_016868166.1 5 574
XM_017012678.1 XP_016868167.1 6 1036
XM_017012679.1 XP_016868168.1 5 1389
XM_017012680.1 XP_016868169.1 5 688
XM_017012681.1 XP_016868170.1 5 551
XM_017012682.1 XP_016868171.1 5 1101
XM_017012683.1 XP_016868172.1 6 885
GenBank mRNA AF282250.1 - 6 655
AK123862.1 - 5 469
AK313054.1 - 5 524
AY007302.1 - 5 582
AY726595.1 - 4 410
BC020200.1 - 5 706 3944302

View Full Product Details