Gene Transcripts

Gene Symbol
ANGPTL6
Entrez Gene ID
83854
Gene Name
angiopoietin like 6
Gene Aliases
AGF, ARP5
Chromosome Location
Chr. 19: 10092337 - 10106194 on Build GRCh38
UniGene ID
Hs.712850
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321411.1 NP_001308340.1 5 1118
NM_031917.2 NP_114123.2 5 1194
XM_005260091.3 XP_005260148.1 5 1361
XM_011528347.2 XP_011526649.1 6 1223
XM_011528348.2 XP_011526650.1 6 1397
XM_011528349.2 XP_011526651.1 7 1856
XM_011528350.2 XP_011526652.1 6 1708
XM_017027347.1 XP_016882836.1 6 1381
GenBank mRNA AB054064.1 - 5 1194
AF230330.1 - 5 1243
AY358276.1 - 5 1119
BC142632.1 - 5 1112 40147717

View Full Product Details