Gene Transcripts

Gene Symbol
ZCCHC7
Entrez Gene ID
84186
Gene Name
zinc finger CCHC-type containing 7
Gene Aliases
AIR1, HSPC086
Chromosome Location
Chr. 9: 37120169 - 37358149 on Build GRCh38
UniGene ID
Hs.654700
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289119.1 NP_001276048.1 3, 4 842
NM_001289120.1 NP_001276049.1 3, 4 865
NM_001289121.1 NP_001276050.1 3, 4 983
NM_032226.2 NP_115602.2 3, 4 838
NR_110317.1 2, 3 207
XM_005251608.4 XP_005251665.1 3, 4 1747
XM_005251612.2 XP_005251669.1 3, 4 805
XM_011518050.1 XP_011516352.1 3, 4 833
XM_011518051.2 XP_011516353.1 2, 3 5770
XM_017015197.1 XP_016870686.1 3, 4 960
XM_017015198.1 XP_016870687.1 3, 4 1446
XM_017015199.1 XP_016870688.1 2, 3 763
XM_017015200.1 XP_016870689.1 2, 3 1036
XM_017015201.1 XP_016870690.1 2, 3 182
GenBank mRNA AK026264.1 - 3, 4 728
AK074608.1 - 2, 3 327
AK303193.1 - 2, 3 116
AK315128.1 - 3, 4 855
BC005990.1 - 3, 4 771
BC036940.1 - 3, 4 956 5399765

View Full Product Details