Gene Transcripts

Gene Symbol
GJA10
Entrez Gene ID
84694
Gene Name
gap junction protein alpha 10
Gene Aliases
CX62
Chromosome Location
Chr. 6: 89894469 - 89896100 on Build GRCh38
UniGene ID
Hs.334499
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032602.1 NP_115991.1 1 1464
GenBank mRNA AF296766.1 - 1 1464

View Full Product Details